Masterflex® Single-Use Transfer Tubing, Gamma-Irradiated C-Flex® Clear, Avantor®
Proveedor: Avantor Fluid Handling
MFLX96124-18EA 424 EUR
MFLX96124-18 MFLX96124-05 MFLX96124-11 MFLX96124-16
Masterflex® Single-Use Transfer Tubing, Gamma-Irradiated C-Flex® Clear, Avantor®
Tuberías Tubo de transferencia
Reduce the risk of microbial contamination.
  • Ideal for general -purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Gamma sensitive labeling confirms treatment
  • Ultra smooth inner surface results in fewer extractables
Masterflex® gamma-irradiated transfer tubing protects your sensitive processes and products from accidentalMore Product Information