Masterflex® Ismatec® Pump Tubing, GORE® STA-PURE® Series PCS Single Elements, Avantor®
Proveedor: Avantor Fluid Handling
MFLX96251-02EA 425 EUR
MFLX96251-02 MFLX96251-06 MFLX96251-08 MFLX96251-16 MFLX96251-26 MFLX96251-28 MFLX96251-30 MFLX96251-34 MFLX96251-40 MFLX96251-42 MFLX96251-58 MFLX96251-62 MFLX96251-04 MFLX96251-10 MFLX96251-12 MFLX96251-20 MFLX96251-24 MFLX96251-32 MFLX96251-36 MFLX96251-48 MFLX96251-50 MFLX96251-52
Masterflex® Ismatec® Pump Tubing, GORE® STA-PURE® Series PCS Single Elements, Avantor®
Tuberías Tubería de bomba peristáltica
Excellent choice for high pressure, low spallation peristaltic tubing applications.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Engineered for long life in peristaltic pump applications
  • Unique composite of expanded PTFE and platinum-cured silicone
Applications: Chemical, criticalMore Product Information