Masterflex® I/P® High-Performance Precision Pump Tubing, Puri-Flex®, Avantor®
Proveedor: Avantor Fluid Handling
MFLX96419-89EA 197 EUR
MFLX96419-89 MFLX96419-70 MFLX96419-88
Masterflex® I/P® High-Performance Precision Pump Tubing, Puri-Flex®, Avantor®
Tuberías Tubería de bomba peristáltica
High-performance and long-life tubing for high-purity applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique application
  • Low spallation when compared to silicone or some