Masterflex® Ismatec® Pump Tubing with Flared Ends, 2-Stop Microbore, Puri-Clear™ LL, Avantor®
Proveedor: Avantor Fluid Handling
MFLX97616-10EA 66.6 EUR
MFLX97616-10 MFLX97616-12 MFLX97616-14 MFLX97616-22 MFLX97616-26 MFLX97616-28 MFLX97616-30 MFLX97616-09 MFLX97616-18 MFLX97616-24
Masterflex® Ismatec® Pump Tubing with Flared Ends, 2-Stop Microbore, Puri-Clear™ LL, Avantor®
Tuberías Tubería de bomba peristáltica
Flared ends for easy extension tubing assembly.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Pthalate-free, USP Class VI PVC
  • Very low Gas Permeability
Two-Stop Flared PVC Color-coded Autoanalysis Tubing for Ismatec® Pumps is USP Class VI tubing that is Phthalate-free. It has a long pumpingMore Product Information