Masterflex® Check Valves, PTFE, Avantor®
Proveedor: Avantor Fluid Handling
98515-14 98515-16 98515-18 98515-12
MFLX98515-14EA 476 EUR
MFLX98515-14 MFLX98515-16 MFLX98515-18 MFLX98515-12
Masterflex® Check Valves, PTFE, Avantor®
Válvulas Válvulas de retención
Use with caustic and high-purity liquids in many applications.
  • Constructed of durable PTFE
Designed for use with corrosive fluids of varying viscosities in chemical, microelectronics, and petroleum industries.