Cleanroom tapes, CR100PC™
Proveedor: MICRONOVA
CR100PC-1LB CR100PC-2LB CR100PC-12DB CR100PC-1DB CR100PC-2DB CR100PC-14WH CR100PC-12WH CR100PC-2WH CR100PC-14YE CR100PC-1YE CR100PC-2YE CR100PC-12DG CR100PC-2DG CR100PC-14RE CR100PC-12RE CR100PC-2RE CR100PC-12TR CR100PC-1TR CR100PC-2TR CR100PC-1WH CR100PC-3DB CR100PC-15DB_U CR100PC-38TR CR100PC-38DB CR100PC-38RE CR100PC-38YE CR100PC-3DG CR100PC-3RE CR100PC-3WH CR100PC-4TR CR100PC-38DG CR100PC-15TR CR100PC-15WH CR100PC-15YE CR100PC-34WH CR100PC-15LB CR100PC-2LG CR100PC-1LG CR100PC-38WH CR100PC-4WH CR100PC-12WHPK CR100PC-15DB CR100PC-1DG CR100PC-1OR CR100PC-1RE CR100-34TRIRM CR100-12WHIRM
MICRCR100PC-1LBEA 963 EUR
MICRCR100PC-1LB MICRCR100PC-2LB 811-0205 MICRCR100PC-1DB MICRCR100PC-2DB MICRCR100PC-14WH MICRCR100PC-12WH MICRCR100PC-2WH MICRCR100PC-14YE MICRCR100PC-1YE MICRCR100PC-2YE MICRCR100PC-12DG MICRCR100PC-2DG MICRCR100PC-14RE MICRCR100PC-12RE MICRCR100PC-2RE MICRCR100PC-12TR MICRCR100PC-1TR MICRCR100PC-2TR 819-0020 MICRCR100PC-3DB MICRCR100PC-15DB_U MICRCR100PC-38TR MICRCR100PC-38DB 811-0206 MICRCR100PC-38YE MICRCR100PC-3DG MICRCR100PC-3RE MICRCR100PC-3WH MICRCR100PC-4TR MICRCR100PC-38DG MICRCR100PC-15TR MICRCR100PC-15WH MICRCR100PC-15YE MICRCR100PC-34WH MICRCR100PC-15LB MICRCR100PC-2LG MICRCR100PC-1LG MICRCR100PC-38WH MICRCR100PC-4WH MICRCR100PC-12WHPK MICRCR100PC-15DB MICRCR100PC-1DG MICRCR100PC-1OR MICRCR100PC-1RE MICRCR100-34TRIRM MICRCR100-12WHIRM
Cleanroom tapes, CR100PC™
Cintas
Extremely conformable, vinyl cleanroom tape with synthetic rubber, clean-peel adhesive; ideal for colour coding, sealing and marking on curved and irregular surfaces in cleanroom maintenance and support areas.
  • Clean peel from wafer cartridges and cleanroom work surfaces
  • Synthetic rubber adhesive ensures a clean peel from most cleanroom surfaces such as polyethylene, plastics and stainless steel
  • Matte finish can be written on with a cleanroom pen
  • Ideal for taping and sealing curved and irregular surfaces
  • Special processing and packaging techniques ensure
More Product Information
Order Now
ESPECIFICACIONES
Environmentally Preferable Sustainable Materials,Sustainable Packaging,Water Efficient,Energy Efficient Sustainable Materials,Sustainable Packaging,Water Efficient,Energy Efficient Sustainable Materials,Sustainable Packaging,Water Efficient,Energy Efficient Sustainable Materials,Sustainable Packaging,Water Efficient,Energy Efficient Sustainable Materials,Sustainable Packaging,Water Efficient,Energy Efficient Sustainable Materials,Sustainable Packaging,Water Efficient,Energy Efficient