Dual plate mop adapter, LyoTool™
Proveedor: MICRONOVA
MZSM56-18 MZSM56-14IR MZSM56-18IR MZSM11-14IR MZSM11-18 MZSM56-14 MZSM11-14 MZSM11-18IR QDBP14-03 QDBP14-02 QDBP14-04 QDBP18-03 MZSM56-14IREO MZSM56-14SIREO
141-1019EA 648 EUR
141-1019 129-0743 141-1020 141-1017 MICRMZSM11-18 129-0742 129-0973 MICRMZSM11-18IR MICRQDBP14-03 MICRQDBP14-02 MICRQDBP14-04 MICRQDBP18-03 MICRMZSM56-14IREO MICRMZSM56-14SIREO
Dual plate mop adapter, LyoTool™
Mopas
A dual-plated mop, specially designed to clean the top and bottom of lyophiliser shelves with one pass. The adjustable, compression fit plates ensure even pressure and consistent contact for optimal sanitisation. Works well on any variety of equipment shelves spaced 3 to 6" apart.
  • Cleans top and bottom of shelves at the same time
  • Spring loaded plates for consistent pressure up and down
  • Front nose guard prevents extending beyond rack
  • Adjust to match shelf height with a simple twist of the knob
  • Use with SnapMop™ and PocketMop™ series flat head mop covers