ProCare Protein Check
Proveedor: MIELE
11626810
MIEL11626810EA 0 EUR
MIEL11626810
ProCare Protein Check
Soluciones de limpieza Desinfectantes
Protein check  for the detection and qualitative assessment of possible protein residues.
  • Test for residues after cleaning and thermal disinfection
  • Precise application
  • Short exposure time of 3 minutes
  • Three pens per packaging unit
  • Up to 50 applications per pen
  • Thermodisinfectors
  • Cleaning and disinfection machines, medicine
  • External dimension: 157×232×28 mm