Amicon® Pro Affinity Concentration Kit, Ultra-0.5 Device
Proveedor: Merck
ACK5100PG ACR5000PG ACK5003GS ACK5003NT ACK5003PA ACK5003PG ACK5010GS ACK5010PA ACK5010PG ACK5030GS ACK5030NT ACK5030PG ACK5050GS ACK5050NT ACK5050PA ACK5050PG ACK5100GS ACK5100NT ACK5100PA ACR5000PA
MILFACK5100PGEA 0 EUR
MILFACK5100PG MILFACR5000PG MILFACK5003GS MILFACK5003NT MILFACK5003PA MILFACK5003PG MILFACK5010GS MILFACK5010PA MILFACK5010PG MILFACK5030GS MILFACK5030NT MILFACK5030PG MILFACK5050GS MILFACK5050NT MILFACK5050PA MILFACK5050PG MILFACK5100GS MILFACK5100NT MILFACK5100PA MILFACR5000PA
Amicon® Pro Affinity Concentration Kit, Ultra-0.5 Device
Filtros Filtros centrífugos
A centrifugal tool for purifying and concentrating His-tagged proteins, GSH-tagged proteins and antibodies.
  • Minimum final concentrate volume: 15 µl
  • Colour code: Clear with pink cap
  • Non sterile
  • Ø×L: 297×120 mm
The device combines affinity-based spin column purification with downstream sample concentration and buffer exchange. Featuring a large reservoir that accommodates a range of sample volumes, the device reduces the need for multiple centrifugation steps. Simply attach the included Amicon® Ultra filter for simultaneous elution, concentration andMore Product Information