Stericup® E-GP Funnel-less Sterile Vacuum Filtration System
Proveedor: Merck
SEGPU0538 SEGPU0545 SEGPU1138 SEGPU1145
MILFSEGPU0538EA 238 EUR
MILFSEGPU0538 MILFSEGPU0545 MILFSEGPU1138 MILFSEGPU1145
Stericup® E-GP Funnel-less Sterile Vacuum Filtration System
Filtros Filtros de la parte superior de botellas
Work with ease and filter with confidence with Stericup® and Steritop® filtration systems. The Stericup® E and Steritop® E sterile filters have been designed to reduce plastic and packaging.
  • The filter collar disconnects from the receiver bottle with just a quarter turn reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood
  • Bold product identification on the filter collar plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection
  • Click