Vented Millex® GS Syringe Filter
Proveedor: Merck
SLGSVR255F
MILFSLGSVR255FEA 325 EUR
MILFSLGSVR255F
Vented Millex® GS Syringe Filter
Filtros Filtros de jeringas
Vented Millex®-GS filter units are syringe filters for sterilisation of low volume aqueous solutions where venting is desired.
  • Clarification of water, buffers or aqueous solutions
  • Mixed Cellulose Esters (MCE)
This single-use product consists of a 0,22 μm hydrophilic MF-MilliporeTM mixed cellulose esters (MCE) membrane filter sealed in a PVC housing with a female Luer-Lok™ inlet connector and a male Luer-slip outlet connector. A 0,03 μm hydrophobic PTFE filter in the dome prevents air locking by automatically venting air introduced upstream.