SNAP i.d.® Starter Packages
Proveedor: Merck
STARTMIDI STARTMINI STARTMM STARTMULTI
MILFSTARTMIDIEA 2130 EUR
MILFSTARTMIDI MILFSTARTMINI MILFSTARTMM MILFSTARTMULTI
SNAP i.d.® Starter Packages
Analizadores Analizadores de proteínas
Western blot packages with unbelievable savings! Save time and money by choosing the package that best serves your needs! The SNAP i.d.® system applies a vacuum to actively drive reagents through the membrane, promoting antigen binding and thorough washing, enabling you to better optimise your Western blotting.
  • Superior: Increased antibody-antigen binding, enhanced washes, and antibody recollection
  • Fast: 30-minute immunodetection
  • Faster testing of different antibodies
  • Higher throughput of Western blots each day
  • Draws reagents through blotting membrane