SNAP i.d. 2.0 Protein Detection Systems Mini
Proveedor: Merck
TISNAP2MINI
MILFTISNAP2MINIEA 0 EUR
MILFTISNAP2MINI
SNAP i.d. 2.0 Protein Detection Systems Mini
Analizadores Analizadores de proteínas
Developed to meet the needs of our Western blotting customers, the SNAP i.d. 2.0 system produces blots of a very high quality. Unique vacuum-driven technology and a built-in flow distributor actively drive reagents through the membrane.
Información para pedidos: This system includes the SNAP i.d. 2.0™ base, two mini blot holding frames, two antibody collection trays, vacuum tubing, rolling pad and blot roller, two wetting trays and a quick start guide.