Milli-Q SQPAK™ TOC Quanta Cartridge
Proveedor: Merck
SQPAKT0CM1
MILLSQPAKT0CM1EA 402 EUR
MILLSQPAKT0CM1
Milli-Q SQPAK™ TOC Quanta Cartridge
Filtros Cartuchos de filtros
The SQPAK™ TOC Quanta cartridge produces ultrapure (Type 1) water at 18,2 MΩ·cm resistivity and TOC ≤5 ppb. The cartridge contains IQnano™ ion-exchange media to remove ions down to trace levels and high-grade synthetic activated carbon that targets traces of organic contaminants.
  • Easy replacement thanks to the Twist and Lock mechanism
  • Need for cartridge replacement is indicated by an orange blinking pictogram light on the system
Ultrapure label and blue colour of the SQPAK™ Quanta cartridge are repeated on the production station interface, to informMore Product Information