Water purification systems, AFS® 8/16/24

Proveedor: Merck
ZAFS0240EU ZAFS0240FR ZAFS0240GE ZAFS0240IT ZAFS0240SP ZAFS0240SZ ZAFS0240UK ZAFS0080FR ZAFS0160EU ZAFS0080BE ZAFS0080EU ZAFS0080GE ZAFS0080IT ZAFS0080SP ZAFS0080SZ ZAFS0080UK ZAFS0160BE ZAFS0080DK ZAFS0160FR ZAFS0160DK ZAFS0160GE ZAFS0160IT ZAFS0160SP ZAFS0160SZ ZAFS0160UK ZAFS0240BE ZAFS0240DK
MILLZAFS0240EUEA 9970 EUR
MILLZAFS0240EU MILLZAFS0240FR MILLZAFS0240GE MILLZAFS0240IT MILLZAFS0240SP MILLZAFS0240SZ MILLZAFS0240UK MILLZAFS0080FR MILLZAFS0160EU MILLZAFS0080BE MILLZAFS0080EU MILLZAFS0080GE MILLZAFS0080IT MILLZAFS0080SP MILLZAFS0080SZ MILLZAFS0080UK MILLZAFS0160BE MILLZAFS0080DK MILLZAFS0160FR MILLZAFS0160DK MILLZAFS0160GE MILLZAFS0160IT MILLZAFS0160SP MILLZAFS0160SZ MILLZAFS0160UK MILLZAFS0240BE MILLZAFS0240DK
Water purification systems, AFS® 8/16/24
Purificación de agua Sistemas de agua pura y ultra pura
Economical water system, producing CLRW (Clinical Laboratory Reagent Water) quality with a dispensing flow rate to analyser up to 2 l/min. For clinical analysers with pure water needs up to 480 L daily

  • Cost-effective: Optimised lifetimes for pretreatment and polishing packs, low feed water use
  • Full monitoring and automatic water quality archiving; information can be stored for up to 6 months
  • Small footprint, allowing installation wherever it’s convenient: on the wall, on or under the bench, or on a cart
  • Intuitive icon-based system display facilitates
More Product Information
Order Now

ESPECIFICACIONES

Modelo AFS® 8 AFS® 16 AFS® 24
Environmentally Preferable
Agua de alimentación Portable tap water at a maximum pressure of 6 bar Portable tap water at a maximum pressure of 6 bar Portable tap water at a maximum pressure of 6 bar
Velocidad de flujo 8 L/h @ 25 °C 16 L/h @ 25 °C 24 L/h @ 25 °C
Contenido TOC <30 ppb <30 ppb <30 ppb
Resistividad a +25 °C >15 MΏ.cm >15 MΏ.cm >15 MΏ.cm
Bacteria < 1 CFU/ml < 1 CFU/ml < 1 CFU/ml
Ancho × Fondo × Alto 268×426×585 mm 268×426×585 mm 268×426×585 mm
Peso <28 kg <28 kg <28 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR