Microarray scanner, GenePix® 4100A
Proveedor: MOLECULAR DEVICES
GENEPIX4100A-U
MLDVGENEPIX4100A-UEA 0 EUR
MLDVGENEPIX4100A-U
Microarray scanner, GenePix® 4100A
Instrumentos Microarray
GenePix® 4100A Microarray Scanner offers its simplicity, reliability and flexibility in microarray-based research, be it in genomics, proteomics, or novel applications. It has all the quality, sensitivity, reliability and ease-of use of more expensive scanners, but in a price range and bench top footprint that makes it ideal for individual lab use.
  • Compact, affordable and easy-to-use
  • Superior imaging accuracy
  • Outstanding reproducibility
  • Flexible fluorophore collection
  • Optimise settings on-the-fly  
  • Greater signal-to-noise ratio
The GenePix® 4More Product Information