Microplate readers, multi-mode, SpectraMax® iD5
Proveedor: MOLECULAR DEVICES
ID5-STD ID5-BL-INJ ID5BLINJSCACA3 ID5-BL ID5-INJ ID5BLSCACAD3 ID5INJSCACAD3 ID5BLINJSCACAD ID5-BL-SC-ACAD ID5INJSCACAD
735-0358EA 0 EUR
735-0358 735-0378 MLDVID5BLINJSCACA3 735-0377 735-0379 MLDVID5BLSCACAD3 MLDVID5INJSCACAD3 MLDVID5BLINJSCACAD MLDVID5-BL-SC-ACAD MLDVID5INJSCACAD
Microplate readers, multi-mode, SpectraMax® iD5
Lectores de microplacas
The SpectraMax® iD5 multi-mode microplate reader is a complete laboratory solution to increase your research capabilities and comes with built-in absorbance, fluorescence, luminescence, time-resolved fluorescence (TRF) and tuneable fluorescence polarisation (FP) read modes. In addition, the SpectraMax® iD5 can be expanded to include bottom-read luminescence, TR-FRET, HTRF®, BRET, dual luciferase reporter assays with injectors and Western blot detection.
  • Easily set up protocols, run experiments, view tutorial videos, and control user access with the large