Cintas de enmascaramiento industriales, 301E
Proveedor: 3M
301E-18MM 301E-24MM 301E-36MM 301E-48MM
MMMA301E-18MMEA 280 EUR
MMMA301E-18MM MMMA301E-24MM MMMA301E-36MM MMMA301E-48MM
Cintas de enmascaramiento industriales, 301E
Cintas
301E masking tape is a thick crepe paper tape for industrial applications that offers good water resistance.
  • Withstands surface temperatures up to 225 °F/107 °C for 30 minutes
  • Resistant to solvents and moisture
  • Easy and neat detachment of many heating systems after curing
  • Designed for indoor use
  • Tensile strength: 95 [Newton/25 mm]
  • Crepe paper back material
It provides good holding power that resists lifting or curling, making it suitable for air drying and oven curing of paint and lacquer.
Using this proprietary solvent-free adhesive, the tapeMore Product Information