Performance box sealing tape 3444, Scotch®
Proveedor: 3M
34445050
MMMA34445050EA 461 EUR
MMMA34445050
Performance box sealing tape 3444, Scotch®
Cintas
Seal cartons by using Scotch® Performance Box Sealing Tape 3444. Our sealing tape is made from non-silicone flat back impregnated paper, coated with a natural rubber adhesive. It sticks to a wide variety of surfaces.
  • Apply to carton sealing applications
  • Use as a single strip closure of lightweight boxes, repaired and miscellaneous packaging
  • Get non-silicone flat-back impregnated paper, coated with a high tack, natural rubber adhesive
  • Good alternative to plastic tapes
  • Good performance in damp and cold shipping and storage environments