Protective coveralls, 4510
Proveedor: 3M
4510S 4510L 4510M 4510XL 4510XXL 45104XL 45103XL
MMMA4510SEA 292 EUR
MMMA4510S MMMA4510L MMMA4510M MMMA4510XL MMMA4510XXL MMMA45104XL MMMA45103XL
Protective coveralls, 4510
Prendas desechables
3M™ Protective coverall 4510 offers economic protection against infective agents, certain hazardous dusts, and limited liquid splashes. The PE laminate includes micro pores which allow heat and moisture to evaporate from the skin and out through the suit, offering a light and comfortable yet protective solution.
  • Type 5/6 Coverall designed to offer cost effective protection against hazardous dusts and limited liquid splashes
  • Made of a soft, lightweight, non-woven microporous laminate for splash protection, strength, and breathability
  • 2-panel hood design, 2-