Protective coveralls, 4520
Proveedor: 3M
4520L 4520M 4520XL 4520XXL 4520S 4520CSL 4520CSXL 4520CSM 4520CSXXL
113-0795EA 280 EUR
113-0795 113-0796 113-0797 113-0798 MMMA4520S MMMA4520CSL MMMA4520CSXL MMMA4520CSM MMMA4520CSXXL
Protective coveralls, 4520
Prendas desechables
3M™ Protective Coverall 4520 has been engineered to offer multiple layers of SMMMS for effective hold-out and strength at a lightweight 43 gsm. Superbly breathable and soft with a cloth-like feel, it can be worn for extended periods comfortably and protects against static discharge, hazardous particulates, and liquid splash.
  • Coveralls protect against certain light liquid splashes and hazardous dusts
  • Anti-static treatment can help reduce static build-up
  • Does not contain natural rubber latex components
  • Seamless shoulders and sleeve tops offer extra strength
More Product Information
Order Now
ESPECIFICACIONES
Modelo 4520
Environmentally Preferable
Antiestático +
Género
Aplicaciones de vestimenta Chemical protection,General Purpose,Laboratory