Chemical Resistant Overalls, 4545
Proveedor: 3M
45452XL 45453XL 45454XL 4545L 4545M 4545XL 4545S
MMMA45452XLEA 383 EUR
MMMA45452XL MMMA45453XL MMMA45454XL MMMA4545L MMMA4545M MMMA4545XL MMMA4545S
Chemical Resistant Overalls, 4545
Prendas desechables
These disposable overalls are made of a laminated microporous material consisting of polyethylene and polyester. The garments provide an excellent barrier against hazardous dusts (type 5) and certain light liquid splashes (type 6). Certified to also protect against radioactive particles and biological agents and reduce build up of static charge.
  • Soft, well-draping material
  • Elasticated waist and ankles for freedom of movement and knitted cuffs for extra comfort
  • Three-panel hood design for a better fit and compatibility with other PPE
  • Two-way zip and