Adflo™ Powered Air Purifying Respirator Systems with Speedglas™ G5-01 Series Welding Helmet
Proveedor: 3M
7100256131 7100257943
MMMA7100256131EA 3140 EUR
MMMA7100256131 MMMA7100257943
Adflo™ Powered Air Purifying Respirator Systems with Speedglas™ G5-01 Series Welding Helmet
Respiradores Sistemas de respiración
The Adflo™ Powered air purifying respirator system with 3M™ Speedglas™ G5-01 series Welding helmet provides all of the benefits of the G5-01 series welding helmet with a convenient Adflo™ Powered air purifying respirator system.
  • Robust, compact design
  • Ergonomic, adjustable head suspension
  • 73×109 mm viewing area
  • Flip-front welding shield allows for a clear, wide-view protective visor for grinding and setup
  • Available in two different auto-darkening lens options (VC/TW)
  • Audible and visual alarms signal low airflow and low battery charge
  • G5 series storage/