Trouble Shooter Aerosol Cleaner
Proveedor: 3M
CHINTENS
MMMACHINTENSEA 337 EUR
MMMACHINTENS
Trouble Shooter Aerosol Cleaner
Soluciones de limpieza Detergentes y Limpiadores de Superficie
Trouble shooter cleaners are cleaning sprays for removing tough stains from difficult-to-reach areas. They easily remove heavy soil, wax, or floor finish build-ups from hard surfaces.
  • Easy-to-apply with the directional nozzle
  • Easily removes stains from water resistant surfaces
  • Clings to vertical surfaces without dripping
  • Designed for use with the 3M™ Doodlebug Cleaning System for best results
Remove tough stains from difficult-to-reach areas with 3M™ Trouble shooter cleaners. The non-ammoniated formula penetrates and loosens stains quickly, meaning noMore Product Information