Powered air respirator visor pivot kits, Versaflo™
Proveedor: 3M
M-960
MMMAM-960EA 81.9 EUR
MMMAM-960
Powered air respirator visor pivot kits, Versaflo™
Respiradores Sistemas de respiración
Versaflo™ Visor Pivot Kit M-960 is a replacement part designed for use with 3M™ Versaflo™ M-Series Headtops.
  • Replacement visor pivot kit
  • Designed for use with 3M™ Versaflo™ M-Series Headtops
Ensure visors are performing correctly with the replacement Versaflo™ Visor Pivot Kit, M-960. This pivot kit is designed to be used as a spare part to replace worn or broken apparatus in Versaflo™ M-Series Headtops.