Powered air respirator spark arrestor for Versaflo™ powered air turbo TR-600 series, Versaflo™
Proveedor: 3M
TR-662
MMMATR-662EA 41.6 EUR
MMMATR-662
Powered air respirator spark arrestor for Versaflo™ powered air turbo TR-600 series, Versaflo™
Respiradores Sistemas de respiración
The Versaflo™ Spark Arrestor TR-662 is a spark arrestor for Versaflo™ Powered Air Turbo TR-600 Series filters. It prevents damage from sparks and other hot, airborne particles.
  • Prevent damage to TR-600 Series filters
  • Must be used whenever sparks or hot, airborne particles are present in the workplace
  • Cleanable for reuse
The spark arrestor must be used whenever sparks or hot, airborne particles are present in the workplace. It can also act as a pre-filter by capturing larger particles which would otherwise be drawn in where they can clog the filter. ItMore Product Information