Powered air respirator airflow indicator TR-971, Versaflo™
Proveedor: 3M
TR-971
MMMATR-971EA 56.9 EUR
MMMATR-971
Powered air respirator airflow indicator TR-971, Versaflo™
Respiradores Sistemas de respiración
Versaflo™ Air Flow Indicator TR-971 is designed to confirm the manufacturer's design minimum air flow rate of Versaflo™ Powered Air Turbo is being exceeded.
  • Testing enabled at various elevations and temperatures
  • Air flow indicator must be tested before using system
  • Easy to use
Ensure a Versaflo™ Powered Air Turbo system is exceeding its minimum design air flow rate with the Versaflo™ Air Flow Indicator, TR-971. This air flow indicator provides a simple and reliable way to check the airflow of a Versaflo™ Turbo unit at the various elevations andMore Product Information