SparkPlugs® Detect Disposable Earplugs
Proveedor: Moldex
7809 7859 785902
111-0721EA 235 EUR
111-0721 111-0723 MOLD785902
SparkPlugs® Detect Disposable Earplugs
Tapones para los oídos Tapones para los oídos, desechables
SparkPlugs® earplugs are made of soft, extra light foam.
  • Tapered shape fits easily in the ear canal and seals gently and snugly without pressure
  • Fully detectable, with metal detectable cord
  • Also available in the hygienic MoldexStation dispenser system
Their bright blue colour makes them perfect for food processing applications.
SNR: 32 dB
Certificaciones: EN 352-2