Anti-IgG Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Cytiva
NA931-100UL
NA931-100ULEA 114.96 EUR
NA931-100UL
Anti-IgG Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Highly species specific HRP-conjugated antibodies optimized for use with Amersham ECL Western Blotting Detection Reagents.

  • Soluble in cold water and hot water

Optimized for use with our range of Amersham ECL Western blotting detection reagents.

The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2 °C to 8 °C.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR