Non-phthalate PVC tubing, Nalgene™
Proveedor: Thermo Fisher Scientific
8701-0010 8701-0020 8701-9020 8701-0030 8701-9030 8701-0060 8701-4060 8701-0070 8701-0080 8701-0090 8701-9090 8701-0100 8701-0110 8701-0120 8701-4120 8701-0130 8701-0140 8701-4140 8701-0160 8701-0180 8701-0190 8701-0200 8701-0260 8701-0310 8701-0350
NALG8701-0010EA 0 EUR
NALG8701-0010 NALG8701-0020 NALG8701-9020 NALG8701-0030 NALG8701-9030 NALG8701-0060 NALG8701-4060 NALG8701-0070 NALG8701-0080 NALG8701-0090 NALG8701-9090 NALG8701-0100 NALG8701-0110 NALG8701-0120 NALG8701-4120 NALG8701-0130 NALG8701-0140 NALG8701-4140 NALG8701-0160 NALG8701-0180 NALG8701-0190 NALG8701-0200 NALG8701-0260 NALG8701-0310 NALG8701-0350
Non-phthalate PVC tubing, Nalgene™
Tuberías Tubo de transferencia
Nalgene lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins, free from ortho-phthalate plasticisers.
  • Compare to Tygon® E-3603 tubing
  • Marked with Int.Ø and regulatory compliance as well as at 305 mm intervals for easy identification and measurement
  • Durometer hardness is shore A, 60
Nalgene™ Non-phthalate PVC tubing is manufactured from the highest-grade resin materials and performs like classic Nalgene™ 180 tubing but is free from orthophthalate DEHP plasticisers. Resists hardening andMore Product Information
Order Now
ESPECIFICACIONES
Modelo NALG8701-0010 NALG8701-0020 NALG8701-9020 NALG8701-0030 NALG8701-9030 NALG8701-0060 NALG8701-4060 NALG8701-0070 NALG8701-0080 NALG8701-0090 NALG8701-9090 NALG8701-0100 NALG8701-0110 NALG8701-0120 NALG8701-4120 NALG8701-0130 NALG8701-0140 NALG8701-4140 NALG8701-0160 NALG8701-0180 NALG8701-0190 NALG8701-0200 NALG8701-0260 NALG8701-0310 NALG8701-0350
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Presión máx. 2,07 bar 3,45 bar 3,45 bar 2,48 bar 2,48 bar 2,07 bar 2,07 bar 2,76 bar 3,17 bar 1,93 bar 1,93 bar 2,48 bar 2,76 bar 1,52 bar 1,52 bar 2,20 bar 2,76 bar 2,76 bar 2,07 bar 1,38 bar 1,79 bar 2,07 bar 1,79 bar 1,52 bar 1,24 bar
Certificación ambiental NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement