Medical absorbent mats
Proveedor: NEW PIG
MATE1110
NEWPMATE1110EA 279 EUR
NEWPMATE1110
Medical absorbent mats
Materiales para contención de derrames
These medical absobent mats are made from a non-wetting top layer to keep the standing surface dry, a super-absorbent central layer and a non-slip backing that grips to the floor. The mats keep floors dry during wet operating procedures.
  • Reduces slips and falls
  • Easily lifted and disposed of after use which reduces clean up time
  • Minimises tripping hazards
  • Reduces clean up time, it can be easily lifted and disposed after use
  • Simple to use, can be cut into individual sizes
For hospital floors, it absorbs spills from procedures like knee and hipMore Product Information