PIG® Original absorbent socks
Proveedor: NEW PIG
404 204 PIG205 PIG209 PIG218 PIG219 PIG220
NEWP404EA 116.75 EUR
NEWP404 NEWP204 NEWPPIG205 NEWPPIG209 NEWPPIG218 NEWPPIG219 NEWPPIG220
PIG® Original absorbent socks
Materiales para contención de derrames
Formable socks hug corners and surround machine bases to absorb machine leaks, contain puddles and help keep workers safe.
  • Tough and economical; good choice for everyday leak protection
  • Contained corncob filler pulls liquids from the floor for maximum absorbency whilst making cleanup quick and easy
  • Polypropylene skin resists tearing; reduces dust and holds in liquid, even when saturated
  • Absorbs and retains oils, coolants, solvents and water
  • Corncob filler is light-weight and economical to handle general, everyday tasks
Can be incinerated after use toMore Product Information