Spatulas, Trulla type
Proveedor: NICKEL ELECTRO
9470
NICK9470EA 88.1 EUR
NICK9470
Spatulas, Trulla type
Espátulas
Stainless steel, PTFE XYLAN® coated. These spatulas feature a PTFE XYLAN® coating which provides a non stick surface to which very few solid or liquid substances will permanently adhere.
  • Stainless steel base: Spatula can withstand extreme temperatures and chemicals
  • PTFE coating provides chemical inertness, purity, low permeability, heat resistance, toughness, and non stick qualities
  • Excellent non wetting properties make cleaning very easy
  • Spatula can undergo steam or chemical sterilisation
Ideal for biology, microbiology and pharmaceutical applications.