Lids, flat for Clifton stirred baths
Proveedor: NICKEL ELECTRO
LD4-14 LD4-22 LD4-8
NICKLD4-14EA 191 EUR
NICKLD4-14 NICKLD4-22 NICKLD4-8
Lids, flat for Clifton stirred baths
Tapas Bath Lids
Stainless steel flat lids.
  • The use of a lid helps reduce evaporation and assists in keeping samples free from contamination
  • Insulated handles