Anti-ATM Rabbit Polyclonal Antibody (DyLight® 550)

Proveedor: Novus Biologicals
NB100-104R
NOVUNB100-104REA 688 EUR
NOVUNB100-104R
Anti-ATM Rabbit Polyclonal Antibody (DyLight® 550)
Anticuerpos
Type: Primary
Antigen: ATM (ataxia telangiectasia mutated)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR