Anti-RAD51C Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2H11/6]

Proveedor: Novus Biologicals
NB100-177PE
NOVUNB100-177PEEA 650 EUR
NOVUNB100-177PE
Anti-RAD51C Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2H11/6]
Anticuerpos
Type: Primary
Antigen: RAD51C (RAD51 paralog C)
Clonality: monoclonal
Clone: 2H11/6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR