Anti-MAP1LC3A Rabbit Polyclonal Antibody (DyLight® 550)

Proveedor: Novus Biologicals
NB100-2220R
NOVUNB100-2220REA 803 EUR
NOVUNB100-2220R
Anti-MAP1LC3A Rabbit Polyclonal Antibody (DyLight® 550)
Anticuerpos
Type: Primary
Antigen: MAP1LC3A (microtubule-associated protein 1 light chain 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR