Anti-PDK1 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NB100-2384 NB100-2384SS
NOVUNB100-2384EA 558 EUR
NOVUNB100-2384 NOVUNB100-2384SS
Anti-PDK1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PDK1 (pyruvate dehydrogenase kinase, isozyme 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR