Anti-CRYAB Rabbit Polyclonal Antibody (FITC (Fluorescein))

Proveedor: Novus Biologicals
NB100-2519F
NOVUNB100-2519FEA 722 EUR
NOVUNB100-2519F
Anti-CRYAB Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticuerpos
Type: Primary
Antigen: CRYAB (crystallin, alpha B)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR