Anti-LOX Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Novus Biologicals
NB100-2527H
NOVUNB100-2527HEA 803 EUR
NOVUNB100-2527H
Anti-LOX Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Type: Primary
Antigen: LOX (lysyl oxidase)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR