Anti-PTPN13/PTPL1 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NB100-56139
NOVUNB100-56139EA 494 EUR
NOVUNB100-56139
Anti-PTPN13/PTPL1 Rabbit Polyclonal Antibody
Anticuerpos
The PTPN13 / PTPL1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PTPN13 / PTPL1. This antibody reacts with human. The PTPN13 / PTPL1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000-1:5000

Type: Primary
Antigen: PTPN13/PTPL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR