Anti-Alas1 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NB100-56415
NOVUNB100-56415EA 444 EUR
NOVUNB100-56415
Anti-Alas1 Rabbit Polyclonal Antibody
Anticuerpos
The Alas1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Alas1. This antibody reacts with human. The Alas1 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry-Paraffin.

Simple Western,

Type: Primary
Antigen: Alas1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR