Anti-TLR7 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NB100-56682
NOVUNB100-56682EA 434 EUR
NOVUNB100-56682
Anti-TLR7 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TLR7 (toll-like receptor 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

ESPECIFICACIONES

Simbolo antígeno TLR7
Nombre del antígeno Toll-like Receptor 7
Environmentally Preferable
Sinónimos de antígenos TLR7-like
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human,Mouse
Huésped Rabbit
ID del gen 51284
Isotipo
Western blot Yes
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR