Anti-MYD88 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NB100-56698
NOVUNB100-56698EA 610 EUR
NOVUNB100-56698
Anti-MYD88 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: MYD88 (myeloid differentiation primary response 88)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR