Anti-LOX propeptide Rabbit Polyclonal Antibody (DyLight 488)

Proveedor: Novus Biologicals
NB110-41568G
NOVUNB110-41568GEA 722 EUR
NOVUNB110-41568G
Anti-LOX propeptide Rabbit Polyclonal Antibody (DyLight 488)
Anticuerpos
Rabbit Polyclonal LOX propeptide Antibody [DyLight 488]. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Primate, Porcine.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:250

Type: Primary
Antigen: LOX propeptide
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR