Anti-SAT1 Rabbit Polyclonal Antibody (DyLight® 488)

Proveedor: Novus Biologicals
NB110-41622G
NOVUNB110-41622GEA 723 EUR
NOVUNB110-41622G
Anti-SAT1 Rabbit Polyclonal Antibody (DyLight® 488)
Anticuerpos
Type: Primary
Antigen: SAT1 (spermidine/spermine N1-acetyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR