Anti-DRD1 Mouse Monoclonal Antibody (DyLight® 650) [clone: SG2-D1a]

Proveedor: Novus Biologicals
NB110-60017C
NOVUNB110-60017CEA 722 EUR
NOVUNB110-60017C
Anti-DRD1 Mouse Monoclonal Antibody (DyLight® 650) [clone: SG2-D1a]
Anticuerpos
Type: Primary
Antigen: DRD1 (dopamine receptor D1)
Clonality: monoclonal
Clone: SG2-D1a
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR