Anti-BAP1 Mouse Monoclonal Antibody (DyLight® 680) [clone: 1G8]

Proveedor: Novus Biologicals
NB110-60521FR
NOVUNB110-60521FREA 722 EUR
NOVUNB110-60521FR
Anti-BAP1 Mouse Monoclonal Antibody (DyLight® 680) [clone: 1G8]
Anticuerpos
Type: Primary
Antigen: BAP1 (BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase))
Clonality: monoclonal
Clone: 1G8
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR