Anti-Napsin-A Mouse Monoclonal Antibody (DyLight® 488) [clone: TMU-Ad02]

Proveedor: Novus Biologicals
NB110-68133G
NOVUNB110-68133GEA 722 EUR
NOVUNB110-68133G
Anti-Napsin-A Mouse Monoclonal Antibody (DyLight® 488) [clone: TMU-Ad02]
Anticuerpos
Type: Primary
Antigen: Napsin-A
Clonality: monoclonal
Clone: TMU-Ad02
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR