Anti-HSD3B1 Mouse Monoclonal Antibody (DyLight® 405) [clone: FDO66Q]

Proveedor: Novus Biologicals
NB110-78644V
NOVUNB110-78644VEA 722 EUR
NOVUNB110-78644V
Anti-HSD3B1 Mouse Monoclonal Antibody (DyLight® 405) [clone: FDO66Q]
Anticuerpos
Type: Primary
Antigen: HSD3B1 (hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1)
Clonality: monoclonal
Clone: FDO66Q
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR